Anti-AADAT Goat Polyclonal Antibody

Lieferant: US Biological
A1374-12B
USBIA1374-12BEA 601 EUR
USBIA1374-12B
Anti-AADAT Goat Polyclonal Antibody
Antikörper
Recommended dilution: ELISA: 1:32000; Western Blot: 1-3?g/ml

Type: Primary
Antigen: AADAT (aminoadipate aminotransferase)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR