Anti-Amodiaquine Mouse Monoclonal Antibody [clone: 6D10]

Lieferant: US Biological
A1460
USBIA1460EA 735 EUR
USBIA1460
Anti-Amodiaquine Mouse Monoclonal Antibody [clone: 6D10]
Antikörper
Recommended dilution: ELISA: 1:4000

Type: Primary
Antigen: Amodiaquine
Clonality: monoclonal
Clone: 6D10
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a Kappa
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR