Anti-Amoxicillin Mouse Monoclonal Antibody [clone: 1.BB.832]

Lieferant: US Biological
A1463-100
USBIA1463-100EA 465 EUR
USBIA1463-100
Anti-Amoxicillin Mouse Monoclonal Antibody [clone: 1.BB.832]
Antikörper
Type: Primary
Antigen: Amoxicillin
Clonality: monoclonal
Clone: 1.BB.832
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR