Anti-Amyloid 1-40, 42 Mouse Monoclonal Antibody [clone: 6D4]

Lieferant: US Biological
A2275-75L
USBIA2275-75LEA 708 EUR
USBIA2275-75L
Anti-Amyloid 1-40, 42 Mouse Monoclonal Antibody [clone: 6D4]
Antikörper
Recommended dilution: ELISA: 0. 05 ?g/ml; Western Blot: 1 ?g/ml; Immunochemistry: 0.1-1 ?g/ml

Type: Primary
Antigen: Amyloid 1-40, 42
Clonality: Monoclonal
Clone: 6D4
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human,
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR