Anti-Apramycin Sheep polyclonal antibody unconjugated

Lieferant: US Biological
A2291
USBIA2291EA 695 EUR
USBIA2291
Anti-Apramycin Sheep polyclonal antibody unconjugated
Antikörper
Recommended dilution: ELISA: 18?g/ml

Type: Primary
Antigen: Apramycin
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Sheep
Isotype:
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR