Anti-ANGEL1 Rabbit Polyclonal Antibody (PE (Phycoerythrin))

Lieferant: US Biological
A2292-10H-PE
USBIA2292-10H-PEEA 884 EUR
USBIA2292-10H-PE
Anti-ANGEL1 Rabbit Polyclonal Antibody (PE (Phycoerythrin))
Antikörper
Recommended dilution: FLISA: 1:1000; Western Blot: 1:100-1:500
Do not freeze!

Type: Primary
Antigen: ANGEL1 (angel homolog 1 (Drosophila))
Clonality: polyclonal
Clone:
Conjugation: PE (Phycoerythrin)
Epitope: C-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR