Anti-ANGPTL3 Mouse Monoclonal Antibody [clone: 7H146]

Lieferant: US Biological
A2293-48E
USBIA2293-48EEA 815 EUR
USBIA2293-48E
Anti-ANGPTL3 Mouse Monoclonal Antibody [clone: 7H146]
Antikörper
Recommended dilution: Western Blot: 1-2?g/ml

Type: Primary
Antigen: ANGPTL3 (angiopoietin-like 3)
Clonality: monoclonal
Clone: 7H146
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR