Anti-ACE2 Rabbit Polyclonal Antibody (FITC (Fluorescein))

Lieferant: US Biological
A2295-02D5FITC
USBIA2295-02D5FITCEA 884 EUR
USBIA2295-02D5FITC
Anti-ACE2 Rabbit Polyclonal Antibody (FITC (Fluorescein))
Antikörper
Type: Primary
Antigen: ACE2 (angiotensin I converting enzyme 2)
Clonality: polyclonal
Clone:
Conjugation: FITC (Fluorescein)
Epitope: C-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR