Anti-ACE2 Rabbit Polyclonal Antibody

Lieferant: US Biological
A2295-02W1
USBIA2295-02W1EA 811 EUR
USBIA2295-02W1
Anti-ACE2 Rabbit Polyclonal Antibody
Antikörper
Recommended dilution: Immunochemistry: 2.5 ?g/ml; Western Blot: 2?g/ml

Type: Primary
Antigen: ACE2 (angiotensin I converting enzyme 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR