Anti-ANLN Goat Polyclonal Antibody

Lieferant: US Biological
A2295-12X3
USBIA2295-12X3EA 809 EUR
USBIA2295-12X3
Anti-ANLN Goat Polyclonal Antibody
Antikörper
Recommended dilution: ELISA: 1:4000; Western Blot: 1 ?g/ml

Type: Primary
Antigen: ANLN (anillin, actin binding protein)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human, Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR