Anti-ANLN Goat Polyclonal Antibody
Lieferant: US Biological
A2295-12Y1
USBIA2295-12Y1EA 640 EUR
USBIA2295-12Y1
Anti-ANLN Goat Polyclonal Antibody
Antikörper
Recommended dilution: ELISA: 1:16000; Western Blot: 1-3?g/ml
Type: Primary
Antigen: ANLN (anillin, actin binding protein)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Dog, Human, Mouse, Rat