Anti-SERPINA1 Sheep Polyclonal Antibody

Lieferant: US Biological
A2298-27B1
USBIA2298-27B1EA 802 EUR
USBIA2298-27B1
Anti-SERPINA1 Sheep Polyclonal Antibody
Antikörper
Type: Primary
Antigen: SERPINA1 (serpin peptidase inhibitor, clade A (alpha-1 antiproteinase, antitrypsin), member 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Sheep
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR