Anti-APRT Rabbit polyclonal antibody unconjugated

Lieferant: US Biological
A2300-15A
USBIA2300-15AEA 649 EUR
USBIA2300-15A
Anti-APRT Rabbit polyclonal antibody unconjugated
Antikörper
Recommended dilution: ELISA: 1:1000; Western Blot: 1:100-1:500

Type: Primary
Antigen: APRT (adenine phosphoribosyltransferase)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR