Anti-APRT Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))

Lieferant: US Biological
A2300-15A-HRP
USBIA2300-15A-HRPEA 884 EUR
USBIA2300-15A-HRP
Anti-APRT Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))
Antikörper
Recommended dilution: ELISA: 1:1000; Western Blot: 1:100-1:500

Type: Primary
Antigen: APRT (adenine phosphoribosyltransferase)
Clonality: polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope: C-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR