Anti-ARTN Rabbit Polyclonal Antibody
Lieferant: US Biological
A3580-06A
USBIA3580-06AEA 640 EUR
USBIA3580-06A
Anti-ARTN Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: ARTN (artemin)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human