Anti-NPPA Mouse Monoclonal Antibody [clone: L11-11-D6-B4]

Lieferant: US Biological
A4150-03A-10
USBIA4150-03A-10EA 439 EUR
USBIA4150-03A-10
Anti-NPPA Mouse Monoclonal Antibody [clone: L11-11-D6-B4]
Antikörper
Type: Primary
Antigen: NPPA (natriuretic peptide A)
Clonality: monoclonal
Clone: L11-11-D6-B4
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR