Anti-NPPA Mouse Monoclonal Antibody [clone: A137-M6-19-B6]

Lieferant: US Biological
A4150-03G-100
USBIA4150-03G-100EA 910 EUR
USBIA4150-03G-100
Anti-NPPA Mouse Monoclonal Antibody [clone: A137-M6-19-B6]
Antikörper
Type: Primary
Antigen: NPPA (natriuretic peptide A)
Clonality: monoclonal
Clone: A137-M6-19-B6
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG3 Kappa
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR