Anti-ATXN3 Rabbit Polyclonal Antibody (Biotin)
Lieferant: US Biological
A4159-08A-BIOT
USBIA4159-08A-BIOTEA 884 EUR
USBIA4159-08A-BIOT
Anti-ATXN3 Rabbit Polyclonal Antibody (Biotin)
Antikörper
Type: Primary
Antigen: ATXN3 (ataxin 3)
Clonality: polyclonal
Clone:
Conjugation: Biotin
Epitope: C-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human