Anti-Bacitracin Sheep polyclonal antibody unconjugated

Lieferant: US Biological
B0003-28
USBIB0003-28EA 763 EUR
USBIB0003-28
Anti-Bacitracin Sheep polyclonal antibody unconjugated
Antikörper
Recommended dilution: ELISA: 0. 625 ?g/ml

Type: Primary
Antigen: Bacitracin
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Sheep
Isotype: IgG
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR