Anti-BCL2L1 Mouse monoclonal antibody unconjugated [clone: 5K203]

Lieferant: US Biological
B0807-14F
USBIB0807-14FEA 443 EUR
USBIB0807-14F
Anti-BCL2L1 Mouse monoclonal antibody unconjugated [clone: 5K203]
Antikörper
Recommended dilution: Immunochemistry: 5-10 ?g/ml; Western Blot: ? 1 ?g/ml

Type: Primary
Antigen: BCL2L1 (BCL2-like 1)
Clonality: monoclonal
Clone: 5K203
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a Kappa
Reactivity: Human, Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR