Anti-Bendazac Sheep polyclonal antibody unconjugated

Lieferant: US Biological
B1037-40A
USBIB1037-40AEA 456 EUR
USBIB1037-40A
Anti-Bendazac Sheep polyclonal antibody unconjugated
Antikörper
Type: Primary
Antigen: Bendazac
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Sheep
Isotype: IgG
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR