Anti-Benzo[a]pyrenediol Epoxide Mouse monoclonal antibody unconjugated [clone: 6A485]

Lieferant: US Biological
B1074
USBIB1074EA 910 EUR
USBIB1074
Anti-Benzo[a]pyrenediol Epoxide Mouse monoclonal antibody unconjugated [clone: 6A485]
Antikörper
Type: Primary
Antigen: Benzo[a]pyrenediol Epoxide
Clonality: monoclonal
Clone: 6A485
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR