Anti-Benzo[a]pyrenediol Epoxide Modified DNA Mouse monoclonal antibody unconjugated [clone: 6A486]

Lieferant: US Biological
B1074-15
USBIB1074-15EA 1010 EUR
USBIB1074-15
Anti-Benzo[a]pyrenediol Epoxide Modified DNA Mouse monoclonal antibody unconjugated [clone: 6A486]
Antikörper
Recommended dilution: Immunochemistry: 1:10

Type: Primary
Antigen: Benzo[a]pyrenediol Epoxide Modified DNA
Clonality: monoclonal
Clone: 6A486
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR