Anti-BHMT Mouse monoclonal antibody unconjugated [clone: 7G21]

Lieferant: US Biological
B1082-12B-100
USBIB1082-12B-100EA 608 EUR
USBIB1082-12B-100
Anti-BHMT Mouse monoclonal antibody unconjugated [clone: 7G21]
Antikörper
Recommended dilution: Western Blot: 1:500-1:1000

Type: Primary
Antigen: BHMT (betaine--homocysteine S-methyltransferase)
Clonality: monoclonal
Clone: 7G21
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a Kappa
Reactivity: Human, Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR