Anti-BHMT Goat polyclonal antibody unconjugated

Lieferant: US Biological
B1082-12F
USBIB1082-12FEA 633 EUR
USBIB1082-12F
Anti-BHMT Goat polyclonal antibody unconjugated
Antikörper
Recommended dilution: ELISA: 1:16000; Western Blot: 1:16000; Immunochemistry: 2.5 ?g/ml

Type: Primary
Antigen: BHMT (betaine--homocysteine S-methyltransferase)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Goat
Isotype:
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR