Anti-BHMT Goat polyclonal antibody unconjugated

Lieferant: US Biological
B1082-13
USBIB1082-13EA 601 EUR
USBIB1082-13
Anti-BHMT Goat polyclonal antibody unconjugated
Antikörper
Recommended dilution: ELISA: 1:16000; Western Blot: 0. 03-0.1 ?g/ml

Type: Primary
Antigen: BHMT (betaine--homocysteine S-methyltransferase)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR