Anti-BHMT Mouse monoclonal antibody unconjugated [clone: 8C11H5]

Lieferant: US Biological
B1082-13A
USBIB1082-13AEA 633 EUR
USBIB1082-13A
Anti-BHMT Mouse monoclonal antibody unconjugated [clone: 8C11H5]
Antikörper
Recommended dilution: Western Blot: 1:500-1:2000; Immunochemistry: 1:200

Type: Primary
Antigen: BHMT (betaine--homocysteine S-methyltransferase)
Clonality: monoclonal
Clone: 8C11H5
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR