Anti-Biotin Goat Polyclonal Antibody (Cy5®)

Lieferant: US Biological
B1750-17E
USBIB1750-17EEA 1080 EUR
USBIB1750-17E
Anti-Biotin Goat Polyclonal Antibody (Cy5®)
Antikörper
Type: Primary
Antigen: Biotin (biotin)
Clonality: polyclonal
Clone:
Conjugation: Cy5®
Epitope:
Host: Goat
Isotype:
Reactivity: Goat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR