Anti-Biotin Goat Polyclonal Antibody (Texas Red®)

Lieferant: US Biological
B1750-35C
USBIB1750-35CEA 489 EUR
USBIB1750-35C
Anti-Biotin Goat Polyclonal Antibody (Texas Red®)
Antikörper
Type: Tag
Antigen: Biotin (biotin)
Clonality: polyclonal
Clone:
Conjugation: Texas Red®
Epitope:
Host: Goat
Isotype: IgG
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR