Anti-Biotin Mouse Monoclonal Antibody [clone: 6F40]

Lieferant: US Biological
B1750-38
USBIB1750-38EA 462 EUR
USBIB1750-38
Anti-Biotin Mouse Monoclonal Antibody [clone: 6F40]
Antikörper
Recommended dilution: ELISA: 1:20000

Type: Tag
Antigen: Biotin (biotin)
Clonality: monoclonal
Clone: 6F40
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a Kappa
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR