Anti-BIRC2 Rabbit Polyclonal Antibody
Lieferant: US Biological
B2049-90F
USBIB2049-90FEA 419 EUR
USBIB2049-90F
Anti-BIRC2 Rabbit Polyclonal Antibody
Antikörper
Recommended dilution: Western Blot: 1:1000-1:2000; Immunochemistry: 1:1000-1:5000; Immunoprecipitation: 1:50-1:200
Type: Primary
Antigen: BIRC2 (baculoviral IAP repeat containing 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human