Anti-DEFB1 Goat polyclonal antibody unconjugated

Lieferant: US Biological
B2469-03
USBIB2469-03EA 460 EUR
USBIB2469-03
Anti-DEFB1 Goat polyclonal antibody unconjugated
Antikörper
Recommended dilution: ELISA: 1:4000

Type: Primary
Antigen: DEFB1 (defensin, beta 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR