Anti-BMP11 Rabbit Polyclonal Antibody (Biotin)

Lieferant: US Biological
B2553-38A-BIOT
USBIB2553-38A-BIOTEA 903 EUR
USBIB2553-38A-BIOT
Anti-BMP11 Rabbit Polyclonal Antibody (Biotin)
Antikörper
Type: Primary
Antigen: BMP11
Clonality: polyclonal
Clone:
Conjugation: Biotin
Epitope: N-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR