Anti-CAMP Rabbit Polyclonal Antibody
Lieferant: US Biological
B2671-01G
USBIB2671-01GEA 755 EUR
USBIB2671-01G
Anti-CAMP Rabbit Polyclonal Antibody
Antikörper
Recommended dilution: Immunochemistry: 1:100
Type: Primary
Antigen: CAMP (cathelicidin antimicrobial peptide)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Rat