Anti-BR Serine/Threonine Protein Kinase 1 Rabbit polyclonal antibody unconjugated

Lieferant: US Biological
B2673-75C
USBIB2673-75CEA 522 EUR
USBIB2673-75C
Anti-BR Serine/Threonine Protein Kinase 1 Rabbit polyclonal antibody unconjugated
Antikörper
Recommended dilution: Western Blot: 0.5-1 ?g/ml; Immunochemistry: 5 ?g/ml

Type: Primary
Antigen: BR Serine/Threonine Protein Kinase 1
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR