Anti-BR Serine/Threonine Protein Kinase 1 Rabbit monoclonal antibody unconjugated [clone: 10E9]
Lieferant: US Biological
B2673-75D
USBIB2673-75DEA 608 EUR
USBIB2673-75D
Anti-BR Serine/Threonine Protein Kinase 1 Rabbit monoclonal antibody unconjugated [clone: 10E9]
Antikörper
Recommended dilution: Western Blot: 1:1000; Immunoprecipitation: 1:50
Type: Primary
Antigen: BR Serine/Threonine Protein Kinase 1
Clonality: monoclonal
Clone: 10000000000
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat