Anti-Brain Natriuretic Peptide, Pro Mouse monoclonal antibody unconjugated [clone: 7E102]

Lieferant: US Biological
B2702-01K
USBIB2702-01KEA 1390 EUR
USBIB2702-01K
Anti-Brain Natriuretic Peptide, Pro Mouse monoclonal antibody unconjugated [clone: 7E102]
Antikörper
Type: Primary
Antigen: Brain Natriuretic Peptide, Pro
Clonality: monoclonal
Clone: 7E+102
Conjugation: unconjugated
Epitope: N-terminal
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR