Anti-CA19-9 Cancer Antigen Mouse monoclonal antibody unconjugated [clone: 3H606]

Lieferant: US Biological
C0075-13D
USBIC0075-13DEA 561 EUR
USBIC0075-13D
Anti-CA19-9 Cancer Antigen Mouse monoclonal antibody unconjugated [clone: 3H606]
Antikörper
Type: Primary
Antigen: CA19-9 Cancer Antigen
Clonality: Monoclonal
Clone: 3H606
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR