Anti-CA19-9 Cancer Antigen Mouse monoclonal antibody unconjugated [clone: 5G17]
Lieferant: US Biological
C0075-31
USBIC0075-31EA 266 EUR
USBIC0075-31
Anti-CA19-9 Cancer Antigen Mouse monoclonal antibody unconjugated [clone: 5G17]
Antikörper
Recommended dilution: Immunohistology: 1:100
Type: Primary
Antigen: CA19-9 Cancer Antigen
Clonality: Monoclonal
Clone: 5G17
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Human