Anti-CA19-9 Cancer Antigen Mouse monoclonal antibody unconjugated [clone: 5G18]

Lieferant: US Biological
C0075-31A
USBIC0075-31AEA 1330 EUR
USBIC0075-31A
Anti-CA19-9 Cancer Antigen Mouse monoclonal antibody unconjugated [clone: 5G18]
Antikörper
Type: Primary
Antigen: CA19-9 Cancer Antigen
Clonality: Monoclonal
Clone: 5G18
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR