Anti-Cancer Antigen CA19-9 Mouse Monoclonal Antibody [clone: 12H3]

Lieferant: US Biological
C0075-31C
USBIC0075-31CEA 776 EUR
USBIC0075-31C
Anti-Cancer Antigen CA19-9 Mouse Monoclonal Antibody [clone: 12H3]
Antikörper
Type: Primary
Antigen: Cancer Antigen CA19-9
Clonality: Monoclonal
Clone: 12H3
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR