Anti-CA50, Cancer Antigen Mouse monoclonal antibody unconjugated [clone: 10g164]

Lieferant: US Biological
C0085-01D
USBIC0085-01DEA 614 EUR
USBIC0085-01D
Anti-CA50, Cancer Antigen Mouse monoclonal antibody unconjugated [clone: 10g164]
Antikörper
Recommended dilution: Immunofluorescence: 10 ?g/ml; ELISA: 0.2ng/ml; Flow Cytometry: 10 ?g/ml

Type: Primary
Antigen: CA50, Cancer Antigen
Clonality: Monoclonal
Clone: 10g164
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG3 Kappa
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR