Anti-CA50, Cancer Antigen Mouse monoclonal antibody unconjugated [clone: 12K257]

Lieferant: US Biological
C0085-01E
USBIC0085-01EEA 614 EUR
USBIC0085-01E
Anti-CA50, Cancer Antigen Mouse monoclonal antibody unconjugated [clone: 12K257]
Antikörper
Recommended dilution: ELISA: 0.2ng/ml

Type: Primary
Antigen: CA50, Cancer Antigen
Clonality: Monoclonal
Clone: 12K257
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG3 Kappa
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR