Anti-CAD Rabbit polyclonal antibody unconjugated

Lieferant: US Biological
C0106-05
USBIC0106-05EA 633 EUR
USBIC0106-05
Anti-CAD Rabbit polyclonal antibody unconjugated
Antikörper
Recommended dilution: Western Blot: 1:500-1:3000; Immunochemistry: 10 ?g/ml

Type: Primary
Antigen: CAD (carbamoyl-phosphate synthetase 2, aspartate transcarbamylase, and dihydroorotase)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR