Anti-Calcium Transport Protein 2 Goat Polyclonal Antibody

Lieferant: US Biological
C0122-02A
USBIC0122-02AEA 640 EUR
USBIC0122-02A
Anti-Calcium Transport Protein 2 Goat Polyclonal Antibody
Antikörper
Recommended dilution: ELISA: 1:16000; Western Blot: 1-3?g/ml

Type: Primary
Antigen: Calcium Transport Protein 2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Bovine, Dog, Human, Mouse, Pig, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR