Anti-Calcyclin Rabbit Polyclonal Antibody

Lieferant: US Biological
C1031-90C
USBIC1031-90CEA 633 EUR
USBIC1031-90C
Anti-Calcyclin Rabbit Polyclonal Antibody
Antikörper
Recommended dilution: Immunochemistry: 1:50

Type: Primary
Antigen: Calcyclin
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR