Anti-CA1 Goat polyclonal antibody unconjugated

Lieferant: US Biological
C1105-09
USBIC1105-09EA 640 EUR
USBIC1105-09
Anti-CA1 Goat polyclonal antibody unconjugated
Antikörper
Recommended dilution: ELISA: 1:64000

Type: Primary
Antigen: CA1 (carbonic anhydrase I)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR