Anti-CA1 Rabbit Polyclonal Antibody (Biotin)

Lieferant: US Biological
C1105-09B
USBIC1105-09BEA 626 EUR
USBIC1105-09B
Anti-CA1 Rabbit Polyclonal Antibody (Biotin)
Antikörper
Type: Primary
Antigen: CA1 (carbonic anhydrase I)
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR