Anti-CARBONIC ANHYDRASE-RELATED PROTEIN Rabbit polyclonal antibody unconjugated

Lieferant: US Biological
C1105-12C
USBIC1105-12CEA 454 EUR
USBIC1105-12C
Anti-CARBONIC ANHYDRASE-RELATED PROTEIN Rabbit polyclonal antibody unconjugated
Antikörper
Recommended dilution: Western Blot: 1:1000; Immunochemistry: 1:1000

Type: Primary
Antigen: CARBONIC ANHYDRASE-RELATED PROTEIN
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR