Anti-CA6 Mouse monoclonal antibody unconjugated [clone: 9H79]

Lieferant: US Biological
C1105-65A
USBIC1105-65AEA 802 EUR
USBIC1105-65A
Anti-CA6 Mouse monoclonal antibody unconjugated [clone: 9H79]
Antikörper
Recommended dilution: Flow Cytometry: 25 ?g/ml; ELISA: 0.5-1.0 ?g/ml

Type: Primary
Antigen: CA6 (carbonic anhydrase VI)
Clonality: Monoclonal
Clone: 9H79
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR